Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300530

Sigma-Aldrich

Anti-HtrA2 (OMI) (N-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-High temperature requirement protein A2, Anti-HtrA2, Anti-Prss25, Anti-Serine proteinase OMI, Omi stress-regulated endoprotease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTRA2(27429)

Related Categories

General description

Insulin-like growth factors (IGFs) stimulate the proliferation and differentiation of a vast number of cell types. The action of the growth factors is mediated and controlled by a complex system of components, including several proteases that cleave the IGF-Binding Proteins. HtrA1 is a 480 aa protein that contains an N-terminus homologous to MAC25 (IGFBP7) with a conserved Kazal-type serine protease inhibitor motif, as well as a C-terminal PDZ domain. Semiquantitative RT-PCR and immunoblot analyses showed an approximately 7-fold increase of PRSS11 in osteoarthritis cartilage compared with controls. HTRA2 is released from mitochondria and inhibits the function of XIAP by direct binding in a way similar to SMAC. Moreover, when overexpressed extramitochondrially, HTRA2 induced atypical cell death, which was neither accompanied by a significant increase in caspase activity nor inhibited by caspase inhibitors, including XIAP. A catalytically inactive mutant of HTRA2, however, did not induce cell death. Suzuki et al. (2001) concluded that HTRA2 is a SMAC-like inhibitor of IAP (inhibitor of apoptosis proteins) activity with a serine protease-dependent cell death-inducing activity.

Immunogen

HTRA2 (61-95)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide at the N-term region of human HtrA2 (OMI).

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service