Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300469

Sigma-Aldrich

Anti-DGKI (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DAG kinase ι, Anti-DGK-ι, Anti-Diacylglycerol kinase, ι, Anti-Diglyceride kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGKI(9162)

General description

DGKI (Diacylglycerol kinase iota) is a small, integral membrane protein belonging to the type IV diacylglycerol kinase subfamily. It is mapped on chromosome 7q32.3-q33. The members of the DGK family consist of a catalytic domain and at least two cysteine-rich regions. In type IV family members, it has four ankyrin repeats and a carboxy terminal PDZ binding domain.
DGKI a member of the type IV diacylglycerol kinase subfamily. Diacylglycerol kinases regulate the intracellular concentration of diacylglycerol through its phosphorylation, producing phosphatidic acid. The specific role of the enzyme encoded by this gene is undetermined, however, it may play a crucial role in the production of phosphatidic acid in the retina or in recessive forms of retinal degeneration.

Immunogen

DGKI (895-930)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human DGKI.

Application

Anti-DGKI (C-term) antibody produced in rabbit is suitable for immunohistochemistry (1:50-1:100), indirect ELISA (1:1000) and western blot (1:100-1:500).

Biochem/physiol Actions

DGKI (Diacylglycerol kinase iota) is mainly involved in the maintenance of intracellular concentration of diacylglycerol by phosphorylating other lipids in addition to DAG. DGKI has been predicted to be a potential gene for tumor suppression in colorectal cancers and mutation in DGKI has been linked to the melanoma.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Role of Chromosomal Instability and Epigenetics in Colorectal Cancers Lacking ??-Catenin/TCF Regulated Transcription
Wael M. Abdel-Rahman, et al.
Gastroenterology Research and Practice (2016)
Matthew K Topham et al.
Biochimica et biophysica acta, 1790(6), 416-424 (2009-04-15)
The mammalian diacylglycerol kinases (DGK) are a group of enzymes having important roles in regulating many biological processes. Both the product and the substrate of these enzymes, i.e. diacylglycerol and phosphatidic acid, are important lipid signalling molecules. Each DGK isoform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service