Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1105926

Sigma-Aldrich

Anti-NEIL2 (276-289) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-NEH2, Anti-NEI2, Anti-Nei endonuclease VIII-like 2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$478.00

$478.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$478.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$478.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt ~37 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEIL2(252969)

Immunogen

synthetic peptide corresponding to amino acids 276-289 of human NEIL2

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service