Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S9125

Sigma-Aldrich

Sodium borohydride

~98%

Synonym(s):

Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

~98%

contains

~0.5% magnesium carbonate as anticaking agent

mp

>300 °C (dec.) (lit.)

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raffaello Mazzaro et al.
ChemistryOpen, 4(3), 268-273 (2015-08-08)
Graphene-metal composites have potential as novel catalysts due to their unique electrical properties. Here, we report the synthesis of a composite material comprised of monodispersed platinum nanoparticles on high-quality graphene obtained by using two different exfoliation techniques. The material, prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service