Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S9005

Sigma-Aldrich

Spleen acetone powder, bovine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

bovine

form

powder

storage temp.

−20°C

General description

An immunosuppressive factor was isolated from spleen acetone powder. It was able to inhibit the formation of lymphoblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of immunosuppressive factors extracted from bovine spleen (lymphoid chalone).
Lenfant, M.
Cell and Tissue Kinetics, 11, Cell Tissue Kinet-Cell Tissue Kinet (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service