Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S8890

Sigma-Aldrich

Sodium bisulfite

ACS reagent

Synonym(s):

Sodium hydrogensulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaHSO3
CAS Number:
Molecular Weight:
104.06
EC Number:
UNSPSC Code:
12352300

grade

ACS reagent

composition

SO2, ≥58.5%

impurities

≤0.005% Insoluble matter

anion traces

chloride (Cl-): ≤0.02%

cation traces

As: ≤1 ppm
Fe: ≤0.002%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].OS([O-])=O

InChI

1S/Na.H2O3S/c;1-4(2)3/h;(H2,1,2,3)/q+1;/p-2

InChI key

DWAQJAXMDSEUJJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Note: This product is usually a mixture of sodium bisulfite and sodium metabisulfite.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service