Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S8567

Sigma-Aldrich

SU 4312

≥98% (HPLC)

Synonym(s):

3-(4-Dimethylaminobenzylidenyl)-2-indolinone, 3-[[(4-Dimethyl­amino)phenyl]methylene]-1,3-dihydro-2H-indol-2-one, E/Z mixture

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$200.00

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$200.00

About This Item

Empirical Formula (Hill Notation):
C17H16N2O
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

orange

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CN(C)c1ccc(cc1)\C=C2\C(=O)Nc3ccccc23

InChI

1S/C17H16N2O/c1-19(2)13-9-7-12(8-10-13)11-15-14-5-3-4-6-16(14)18-17(15)20/h3-11H,1-2H3,(H,18,20)/b15-11+

InChI key

UAKWLVYMKBWHMX-RVDMUPIBSA-N

Gene Information

human ... FLT1(2321)

Application

SU 4312 was used to study angiogenesis during development of zebrafish embryos.2

Biochem/physiol Actions

SU 4312 inhibits the neuronal NOS and exhibits neuroprotection against NO-mediated neurotoxicity. It might be effective in treatment of neurodegenerative disorders.1
SU 4312 is a vascular endothelial growth factor (VEGF) receptor protein tyrosine kinase 1/2 and platelet derived growth factor (PDGF) receptor inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

This product is a mixture of the cis- and trans- isomers. The ratio is about 55:45 based on HPLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Vogt et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 238(3), 656-663 (2009-02-25)
Presently, the zebrafish is the only vertebrate model compatible with contemporary paradigms of drug discovery. Zebrafish embryos are amenable to automation necessary for high-throughput chemical screens, and optical transparency makes them potentially suited for image-based screening. However, the lack of
Wei Cui et al.
British journal of pharmacology, 168(5), 1201-1214 (2012-10-16)
SU4312, a potent and selective inhibitor of VEGF receptor-2 (VEGFR-2), has been designed to treat cancer. Recent studies have suggested that SU4312 can also be useful in treating neurodegenerative disorders. In this study, we assessed neuroprotection by SU4312 against 1-methyl-4-phenylpyridinium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service