Skip to Content
MilliporeSigma
All Photos(1)

Documents

S8067

Sigma-Aldrich

Anti-phospho-Synapsin I (pSer9) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Xenopus, human, chicken, rat, mouse

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:1,000
indirect immunofluorescence: 1:1,000
western blot: 1:1,000 using rat brain hipoccampal tissue homogenate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SYN1(6853)
mouse ... Syn1(20964)
rat ... Syn1(24949)

Immunogen

synthetic phosphorylated peptide derived from the region of rat synapsin 1 that is phosphorylated on serine 9.

Physical form

Solution in 10 mM HEPES buffer, pH 7.5, and 150 mM NaCl containing 100 μg/ml bovine serum albumin and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Du et al.
Aging cell, 20(5), e13368-e13368 (2021-05-06)
Mitochondrial dysfunction is one of the early pathological features of Alzheimer's disease (AD). Accumulation of cerebral and mitochondrial Aβ links to mitochondrial and synaptic toxicity. We have previously demonstrated the mechanism by which presequence peptidase (PITRM1)-mediated clearance of mitochondrial Aβ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service