Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7277

Sigma-Aldrich

Sodium bicarbonate

for molecular biology, 99.7-100.3%

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

for molecular biology

Assay

99.7-100.3%

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

density

2.16 g/mL at 25 °C (lit.)

anion traces

sulfur compounds (as SO42-): ≤0.003%

cation traces

Fe: ≤0.001%
K: ≤0.005%
heavy metals (as Pb): ≤5 ppm

foreign activity

DNase, RNase and protease, none detected

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shay P McGuinness et al.
Critical care medicine, 41(7), 1599-1607 (2013-05-21)
Cardiac surgery-associated acute kidney injury occurs in up to 50% of patients and is associated with increased mortality and morbidity. This study aimed to discover if perioperative urinary alkalinization with sodium bicarbonate infusion reduces the prevalence of cardiac surgery-associated acute
Sophia Zoungas et al.
Annals of internal medicine, 151(9), 631-638 (2009-11-04)
Intravenous sodium bicarbonate has been proposed to reduce the risk for contrast-induced nephropathy (CIN). To determine the effect of sodium bicarbonate on the risk for CIN. MEDLINE, PubMed, EMBASE, and the Cochrane Central Register of Controlled Trials from 1950 to
Eric A J Hoste et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 25(3), 747-758 (2009-08-26)
There have been conflicting reports on the use of intravenous administration of sodium bicarbonate for prevention of contrast-induced acute kidney injury (CI-AKI). The aim of this study was to evaluate the use of sodium bicarbonate for prevention of CI-AKI. This
Joshua C Tremblay et al.
The Journal of physiology, 593(3), 723-737 (2014-11-25)
Blood flow through intrapulmonary arteriovenous anastomoses (IPAVA) is increased by acute hypoxia during rest by unknown mechanisms. Oral administration of acetazolamide blunts the pulmonary vascular pressure response to acute hypoxia, thus permitting the observation of IPAVA blood flow with minimal
Nina Dupuis et al.
Human molecular genetics, 24(10), 2771-2783 (2015-02-06)
Dymeclin is a Golgi-associated protein whose deficiency causes Dyggve-Melchior-Clausen syndrome (DMC, MIM #223800), a rare recessively inherited spondyloepimetaphyseal dysplasia consistently associated with postnatal microcephaly and intellectual disability. While the skeletal phenotype of DMC patients has been extensively described, very little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service