All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H37NO2
CAS Number:
Molecular Weight:
299.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCC\C=C\[C@H](O)[C@H](N)CO
InChI
1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h14-15,17-18,20-21H,2-13,16,19H2,1H3/b15-14+/t17-,18+/m1/s1
InChI key
WWUZIQQURGPMPG-MCXRAWCPSA-N
Other Notes
Inactive isomer of sphingosine
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service