Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6573

Sigma-Aldrich

STK3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KRS1, MST2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

235-319 nmol/min·mg

mol wt

~87 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STK3(6788)

Biochem/physiol Actions

STK3, also known as MST2, encodes a protein of 491-amino acid which contains an N-terminal catalytic domain characteristic of STKs.STK3 and STK4 share 94% amino acid identity in the catalytic domain and 78% identity overall. RAF1 has been shown to counteract apoptosis by suppressing the activation of mammalian sterile 20-like kinase (MST2). STK3 is a kinase that is activated by the proapoptotic agents, straurosporine and FAS ligand.STK3 activation presumably allows cells to resist unfavorable environmental conditions.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yavuz Kulaberoglu et al.
Nature communications, 8(1), 695-695 (2017-09-28)
The Hippo tumor suppressor pathway is essential for development and tissue growth control, encompassing a core cassette consisting of the Hippo (MST1/2), Warts (LATS1/2), and Tricornered (NDR1/2) kinases together with MOB1 as an important signaling adaptor. However, it remains unclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service