Skip to Content
MilliporeSigma
All Photos(1)

Documents

S6382

Sigma-Aldrich

trans-Stilbene

~95%

Synonym(s):

trans-1,2-Diphenylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHC6H5
CAS Number:
Molecular Weight:
180.25
Beilstein:
1616740
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

~95%

bp

305-307 °C/744 mmHg (lit.)

mp

123-125 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

c1ccc(cc1)\C=C\c2ccccc2

InChI

1S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12H/b12-11+

InChI key

PJANXHGTPQOBST-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhang et al.
Bioorganic & medicinal chemistry letters, 21(3), 1032-1035 (2011-01-11)
A series of methoxystilbenes (E and Z isomers) related to resveratrol were investigated for their effects on NQO1 induction in murine hepatoma cells and growth inhibitory effects on human cancer cell lines. Both activities were enhanced in compounds with methoxy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service