Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5883

Sigma-Aldrich

Strophanthidol

Synonym(s):

3β,5,14,19-Tetrahydroxy-5β,20(22)-cardenolide, 5β,20(22)-Cardenolide-3β,5,14,19-tetrol, K-Strophanthidol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O6
CAS Number:
Molecular Weight:
406.51
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC12CCC3C(CCC4(O)CC(O)CCC34CO)C1(O)CCC2C5=CC(=O)OC5

InChI

1S/C23H34O6/c1-20-6-3-17-18(4-8-22(27)11-15(25)2-7-21(17,22)13-24)23(20,28)9-5-16(20)14-10-19(26)29-12-14/h10,15-18,24-25,27-28H,2-9,11-13H2,1H3

InChI key

ZNDMLUUNNNHNKC-UHFFFAOYSA-N

General description

Reduced aglycone of K-strophanthin and cymarin, one of the primary excretion products after use of the glycosides in treatment of congestive heart failure.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service