Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5564

Sigma-Aldrich

SRC N1 human

≥60% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$426.00

$426.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$426.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

$426.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected insect cells

Quality Level

form

buffered aqueous glycerol solution

mol wt

56 kDa

concentration

≥60% (SDS-PAGE)
0.3 mg/mL protein (Bradford assay)

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SRC(6714)

Biochem/physiol Actions

Src N1 is a membrane-associated protein tyrosine kinase (PTK).

Physical form

Solution in 50 mM Tris, pH 7.5, 0.05 mM EDTA, 1 mM DTT, 100 mM NaCl, 0.05% NP-40, and 50% glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Chan et al.
Molecular and cellular biology, 17(8), 4667-4676 (1997-08-01)
The neural cell-specific N1 exon of the c-src pre-mRNA is both negatively regulated in nonneural cells and positively regulated in neurons. We previously identified conserved intronic elements flanking N1 that direct the repression of N1 splicing in a nonneural HeLa

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service