Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5139

Sigma-Aldrich

Sulfur trioxide triethylamine complex

Synonym(s):

NSC 68185, NSC 8168, Triethylamine sulfur trioxide complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)3N · SO3
CAS Number:
Molecular Weight:
181.25
Beilstein:
3993165
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

reaction suitability

reagent type: oxidant

mp

~85 °C

storage temp.

2-8°C

SMILES string

O=S(=O)=O.CCN(CC)CC

InChI

1S/C6H15N.O3S/c1-4-7(5-2)6-3;1-4(2)3/h4-6H2,1-3H3;

InChI key

YYHPEVZFVMVUNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for reagent for synthesis of:
  • Anticoagulant and antiplatelet persulfated glycosides or flavonoids containing an oligopolysulfated moiety
  • Narciclasine derivatives for use as anticancer agents
  • Sulfate-conjugated methylprednisolone for use as a colon-specific prodrug
  • Tetrasaccharide substrates of heparanase by glycosylation of disaccharides
  • Synthetic heparin oligosaccharide microarrays for analysis of heparin-protein interactions
  • Dexamethasone 21-sulfate sodium for use as a colon-specific prodrug

replaced by

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service