Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5009

SAFC

[D-Pro4, D-Trp7,9]-substance fragment 4-11

≥97% (HPLC)

Synonym(s):

D-Pro-Gln-Gln-D-Trp-Phe-D-Trp-Leu-Met-NH2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Caranikas et al.
Journal of medicinal chemistry, 25(11), 1313-1316 (1982-11-01)
Several substance P analogues containing various D-amino acid modifications have been synthesized by the solid-phase procedure, detached from the solid support by ammonolysis, and purified by gel filtration combined with reversed-phase chromatography. Three compounds were fair to very potent competitive
Antagonists of substance P.
S Caranikas et al.
European journal of pharmacology, 77(2-3), 205-206 (1982-01-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service