Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5001

Sigma-Aldrich

Butyl stearate

~99%

Synonym(s):

Stearic acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COO(CH2)3CH3
CAS Number:
Molecular Weight:
340.58
Beilstein:
1792866
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

density

0.861 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCCCC

InChI

1S/C22H44O2/c1-3-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22(23)24-21-6-4-2/h3-21H2,1-2H3

InChI key

ULBTUVJTXULMLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical form

Waxy solid below 25°C, clear liquid above 25°C

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple assay for local heating by optical tweezers.
S C Kuo
Methods in cell biology, 55, 43-45 (1997-11-14)
M A Correa et al.
Colloids and surfaces. B, Biointerfaces, 43(2), 108-114 (2005-05-28)
Microemulsions (ME) containing hexadecyltrimethylammonium bromide (HTAB)/ethanol as surfactant, isopropylmyristate (IM) or butylstearate (BS) as oil phase and aqueous buffer were studied. Pseudo-ternary phase diagrams of the investigated systems were obtained at constant surfactant/cosurfactant molar ratio (1:5) by titration in order
W D Bieber et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 22(9), 737-742 (1984-09-01)
The transfer of stearyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate from high- and low-density polyethylenes and polypropylene as well as that of n-butyl stearate from high-impact polystyrene into several foodstuffs and into different food simulants under normal storage conditions was determined radioanalytically. Additionally, the transfer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service