S4939
N-Succinyl-Leu-Leu-Val-Tyr-7-amido-4-trifluoromethylcoumarin
proteasome substrate, ≥98% (HPLC), solid
Select a Size
$931.00
Select a Size
About This Item
$931.00
Recommended Products
Product Name
N-Succinyl-Leu-Leu-Val-Tyr-7-amido-4-trifluoromethylcoumarin, ≥98% (HPLC), solid
Quality Level
Assay
≥98% (HPLC)
form
solid
solubility
DMF: 1 mg/mL, clear, colorless to light yellow
storage temp.
−20°C
SMILES string
CC(C)C[C@H](NC(=O)CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)Nc2ccc3c(OC(=O)C=C3C(F)(F)F)c2
InChI
1S/C40H50F3N5O10/c1-20(2)15-28(45-32(50)13-14-33(51)52)37(55)46-29(16-21(3)4)38(56)48-35(22(5)6)39(57)47-30(17-23-7-10-25(49)11-8-23)36(54)44-24-9-12-26-27(40(41,42)43)19-34(53)58-31(26)18-24/h7-12,18-22,28-30,35,49H,13-17H2,1-6H3,(H,44,54)(H,45,50)(H,46,55)(H,47,57)(H,48,56)(H,51,52)/t28-,29-,30-,35-/m0/s1
InChI key
ZSJPCNKJLSJEQF-DWNBPJMTSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service