Documents
S4642
Wheaton staining dishes
for 20 slides, dishes only
Synonym(s):
900201
About This Item
UNSPSC Code:
41121812
NACRES:
MA.01
material
soda-lime glass
packaging
pack of 3 ea
manufacturer/tradename
WHEATON® 900201
dish capacity
3 x 1 in. (75 x 25 mm) , 10 single slides
3 x 1-1/2 in. (75 x 38 mm) , 19 slides arranged alternately straight across and diagonally
3 x 2 in. (75 x 51 mm) , 20 slides back-to-back of standard size
inside L × W × D
95 mm × 76 mm × 64 mm
General description
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
Features and Benefits
• Manufactured from soda-lime glass• This 20-slide unit is the standard for manual staining procedures• The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size: 3” x 1” (75 x 25mm), 3” x 1-1/2” (75 x 38mm) and 3” x 2” (75 x 51mm) sizes• Approximate inside bottom dimensions (mm): 91 L x 71 W x 60 D
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.