Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

S4139

Sigma-Aldrich

Spermidine

BioReagent, suitable for cell culture

Synonym(s):

1,8-Diamino-4-azaoctane, N-(3-Aminopropyl)-1,4-diaminobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3NH(CH2)4NH2
CAS Number:
Molecular Weight:
145.25
Beilstein:
1698591
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

Assay

≥98% (GC)

form

solid

technique(s)

cell culture | mammalian: suitable

color

clear colorless to light yellow-white

refractive index

n20/D 1.479 (lit.)

density

0.925 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

NCCCCNCCCN

InChI

1S/C7H19N3/c8-4-1-2-6-10-7-3-5-9/h10H,1-9H2

InChI key

ATHGHQPFGPMSJY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; Stimulates T4 polynucleotide kinase activity.
Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.
Spermidine together with putrescine and spermine compose a family of biogenic polyamines (polycations) that are required for the survival of the vast majority of living cells. Polyamines interact with negatively charged molecules such as proteoglycan, glycated proteins and nucleic acids (DNA and RNA). Biogenic polyamines are found to modulate protein synthesis at different levels. This effect may be explained by the ability of polyamines to bind and influence the secondary structure of tRNA, mRNA, and rRNA. Spermine also helps stabilize nucleic acid helical structure and the conformation of glycated proteins such as the histones. Spermine and spermidine are components of various nucleic acid transfection protocols. Spermidine Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.

Spermidine is biogenic polyamine formed from putrescine, a precursor of spermine. It was first detected in human sperm, but occurs widely in nature. It is essential in both normal and neoplastic tissue growth. Spermidine has a role in cell growth processes and the formation and interconversion of spermidine in mammalian cells has been reported. It has been studied in the regulation of tRNA methyltransferase activity and stimulates T4 polynucleotide kinase activity.

Caution

Solutions should be sterile-filtered, not autoclaved, if sterile solution is necessary. Spermidine deaminates with time; solutions should be stored frozen. Prepare new solutions frequently.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marco Prunotto et al.
Laboratory investigation; a journal of technical methods and pathology, 90(6), 929-939 (2010-03-10)
Epithelial-to-mesenchymal transition (EMT) is involved in embryonic development as well as in several pathological conditions. Literature indicates that polyamine availability may affect transcription of c-myc, matrix metalloproteinase (MMP)1, MMP2, TGFbeta(1), and collagen type I mRNA. The aim of this study
Eugenia Morselli et al.
Aging, 1(12), 961-970 (2010-02-17)
Although autophagy has widely been conceived as a self-destructive mechanism that causes cell death, accumulating evidence suggests that autophagy usually mediates cytoprotection, thereby avoiding the apoptotic or necrotic demise of stressed cells. Recent evidence produced by our groups demonstrates that
Varun K Gupta et al.
Nature neuroscience, 16(10), 1453-1460 (2013-09-03)
Age-dependent memory impairment is known to occur in several organisms, including Drosophila, mouse and human. However, the fundamental cellular mechanisms that underlie these impairments are still poorly understood, effectively hampering the development of pharmacological strategies to treat the condition. Polyamines
A Raina et al.
Medical biology, 53(3), 121-147 (1975-06-01)
The biochemistry and biological function of the naturally occurring polyamines, putrescine, spermidine, and spermine, have been reviewed with special reference to animal organisms. These compounds are universally distributed in all living material. Their biosynthesis from ornithine and methionine is accurately
Anita Nielsen et al.
Marine drugs, 10(11), 2584-2595 (2012-12-04)
Staphylococcus aureus is a serious human pathogen that employs a number of virulence factors as part of its pathogenesis. The purpose of the present study was to explore marine bacteria as a source of compounds that modulate virulence gene expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service