Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3321

Sigma-Aldrich

SUN B8155

≥97% (HPLC), solid

Synonym(s):

5-[1-[(2-Aminophenyl)imino]ethyl]-1,6-dihydroxy-4-methyl-2(1H)-pyridinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N3O3
CAS Number:
Molecular Weight:
273.29
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

Assay

≥97% (HPLC)

form

solid

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CC1=CC(=O)N(O)C(O)=C1\C(C)=N\c2ccccc2N

InChI

1S/C14H15N3O3/c1-8-7-12(18)17(20)14(19)13(8)9(2)16-11-6-4-3-5-10(11)15/h3-7,19-20H,15H2,1-2H3/b16-9+

InChI key

SGJLINMLDNLWOS-CXUHLZMHSA-N

Application

SUN B8155 may be used to study calcitonin-mediated cell signaling.

Biochem/physiol Actions

SUN B8155 is a nonpeptide calcitonin receptor agonist.
SUN B8155 is a pyridine derivative that mimics calcitonin, a peptide hormone crucial in maintain the bone homeostasis. It stimulated the formation of cAMP in T47D and UMR106-06 cells expressing calcitonin receptor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Katayama et al.
Biochimica et biophysica acta, 1526(2), 183-190 (2001-04-28)
Calcitonin (CT), a 32-amino acid peptide hormone secreted mainly from the thyroid gland, plays an important role in maintaining bone homeostasis. To discover non-peptide small molecules with biological actions similar to those of CT, a cell-based screening of an in-house

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service