Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3171

Sigma-Aldrich

Sodium chlorate

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO3
CAS Number:
Molecular Weight:
106.44
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

Assay

≥99.0%

impurities

≤0.001% Nitrogen compounds (as N)
≤0.005% Insoluble matter

mp

248-261 °C (lit.)

anion traces

bromate (BrO3-): ≤0.015%
chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.001%

cation traces

Ca, Mg and R2O3 precipitate: ≤0.005%
Fe: ≤5 ppm
K: ≤0.01%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].[O-]Cl(=O)=O

InChI

1S/ClHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

YZHUMGUJCQRKBT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

sulfation biosynthesis inhibitor

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Troeberg et al.
Matrix biology : journal of the International Society for Matrix Biology, 28(8), 463-469 (2009-08-01)
We investigated whether the affinity of tissue inhibitor of metalloproteinases (TIMP)-3 for adamalysins with thrombospondin motifs (ADAMTS)-4 and ADAMTS-5 is affected by the non-catalytic ancillary domains of the enzymes. For this purpose, we first established a novel method of purifying
Simran Kaur et al.
Journal of food protection, 78(9), 1708-1718 (2015-09-01)
Previous studies show that treatment of cantaloupes with chlorine dioxide (ClO2) gas at 5 mg/liter for 10 min results in a significant reduction (P < 0.05) in initial microflora, an increase in shelf life without any alteration in color, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service