S3136
[pGlu6]-Substance P Fragment 6-11
≥95% (HPLC)
Synonym(s):
pGlu-Phe-Phe-Gly-Leu-Met-NH2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
≥95% (HPLC)
UniProt accession no.
storage temp.
−20°C
SMILES string
CSCCC(NC(=O)C(CC(C)C)NC(=O)CNC(=O)C(Cc1ccccc1)NC(=O)C(Cc2ccccc2)NC(=O)C3CCC(=O)N3)C(N)=O
Gene Information
human ... TAC1(6863)
Biochem/physiol Actions
Substance P agonist; increased blood pressure when injected into cerebral ventricles of rat.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Peptides, 5(1), 85-89 (1984-01-01)
Substance P (SP) significantly reduced fighting in mice made aggressive by prolonged isolation. The N-terminal heptapeptide fragment SP (1-7) also reduced fighting. The C-terminal fragment SP(4-11) was without activity, while the shorter C-terminal fragment analog less than E-SP(7-11) significantly increased
The International journal of biochemistry, 19(12), 1233-1236 (1987-01-01)
1. Peptidase(s) activity of nuclear and synaptosomal fraction from cortex and hippocampus of rat brain against pyroGlu6[125I-Tyr8]SP6-11 was evaluated in different concentration of Ca2+, Mg2+, K+ and Na+ in about "isotonic" conditions. 2. The effects of studied ions on the
Neuropeptides, 16(1), 41-49 (1990-05-01)
Five synthetic N-methylated analogs (II-V) of the C-terminal hexapeptide analog of substance P (SP), [pGlu6]SP6-11 (I) were evaluated for their metabolic stability and in vitro spasmogenic activity. The metabolic resistance of the analogs was tested by two SP degrading systems
Biochimica et biophysica acta, 798(1), 28-36 (1984-03-22)
Inactivation of substance P and its C-terminal hexapeptide analog [p-Glu6]substance P6-11 was studied in rat parotid and hypothalamic slices. It was found that in the parotid slice system the decay of substance P induced K+ release occurs concurrently with a
Japanese journal of pharmacology, 58(3), 325-328 (1992-03-01)
The sialogogic response of submandibular glands to analogs of the C-terminal hexapeptide of substance P with various amino acids at the N-terminus was investigated in urethane-anesthetized rats. The rank order of potencies was as follows: SP greater than (pGlu6)SP6-11 much
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service