Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3066

Sigma-Aldrich

SKF-89626 hydrobromide

>98% (HPLC), solid

Synonym(s):

4-(3,4-Dihydroxyphenyl)-4,5,6,7-tetrahydro­thieno[2,3-c]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO2S · HBr
Molecular Weight:
328.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

form

solid

color

white

solubility

H2O: soluble 11 mg/mL

SMILES string

Br[H].Oc1ccc(cc1O)C2CNCc3sccc23

InChI

1S/C13H13NO2S.BrH/c15-11-2-1-8(5-12(11)16)10-6-14-7-13-9(10)3-4-17-13;/h1-5,10,14-16H,6-7H2;1H

InChI key

QAIGGSDECIJPNW-UHFFFAOYSA-N

Gene Information

human ... DRD1(1812)

Application

SKF-89626 hydrobromide may be used in dopaminergic cell signaling studies.

Biochem/physiol Actions

D1 dopamine receptor agonist.
SKF-89626 is an isoquinoline derivative that fully antagonizes dopamine D1 receptor and stimulates adenylate cyclase and modulates dopaminergic-mediated cell responses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E B Nielsen et al.
European journal of pharmacology, 160(2), 253-262 (1989-01-31)
Dopamine (DA) D-1 and D-2 receptor agonists and antagonists were characterized in receptor binding and adenylate cyclase assays with respect to affinity, selectivity and efficacy. The ability of the ligands to interact with the discriminative stimulus effects of d-amphetamine (AMPH)
P H Andersen et al.
European journal of pharmacology, 188(6), 335-347 (1990-06-12)
Dopamine receptor selectivity was investigated for a number of dopamine receptor agonists. In vitro, the benzazepine derivatives, e.g., SKF 38393 and SKF 75670 as well as the isoquinoline derivatives, SKF 89626 and SKF 89615, were D1 receptor-selective. All other compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service