Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S2070

Sigma-Aldrich

Anti-SF4 antibody produced in rabbit

affinity isolated antibody, ammonium sulfate suspension

Synonym(s):

Anti-Splicing Factor 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 72.4 kDa

species reactivity

human, rodent

technique(s)

immunohistochemistry: suitable
western blot: suitable

storage temp.

−20°C

Gene Information

human ... SF4(57794)
mouse ... Sf4(70616)
rat ... Sf4(290666)

Immunogen

synthetic peptide corresponding to human SF4 (amino acids 103-105), conjugated to BSA.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service