Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1761

Sigma-Aldrich

Substance P Fragment 1-9

≥97% (HPLC)

Synonym(s):

Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H77N15O12
CAS Number:
Molecular Weight:
1104.26
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Braunstein et al.
The American review of respiratory disease, 144(3 Pt 1), 630-635 (1991-09-01)
Our purpose was to characterize the tachykinin receptor type involved in nasal obstruction to exogenous substance P in rhinitic patients. We also attempted to assess biochemical and cellular events associated with this response. Nasal challenges were performed in seven patients
J W Growcott et al.
European journal of pharmacology, 84(1-2), 107-109 (1982-10-15)
The effects of substance P nonapeptide amide on inactivation of substance P by the guinea-pig ileum and urinary bladder preparations and of substance P hexapeptide amide, bradykinin and acetylcholine by the guinea-pig urinary bladder, have been investigated. Significant inhibition of
Effect of substance P on the expression of an adhesion molecule ICAM-1 in human vascular endothelial cells.
N Nakagawa et al.
Regulatory peptides, 46(1-2), 223-224 (1993-07-02)
Liang, T., et al.
The Journal of Neuroscience, 10, 1133-1133 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service