Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

S1250

Sigma-Aldrich

(−)-α-Santonin

≥99%

Synonym(s):

(3S,5aS,9bS)-3a,5,5a,9b-Tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)dione, Semenen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18O3
CAS Number:
Molecular Weight:
246.30
Beilstein:
89489
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥99%

mp

172-173 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O

InChI

1S/C15H18O3/c1-8-10-4-6-15(3)7-5-11(16)9(2)12(15)13(10)18-14(8)17/h5,7-8,10,13H,4,6H2,1-3H3/t8-,10-,13-,15-/m0/s1

InChI key

XJHDMGJURBVLLE-BOCCBSBMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-α-Santonin is a sesquiterpene lactone. It is found in the genus Artemisia.[1]

Application

(−)-α-Santonin has been used as a eudesmane-type sesquiterpene to study its effects on impairment of 231MFP breast cancer cell survival.[2]

Biochem/physiol Actions

(−)-α-Santonin exhibits anti-helminthic properteis.[1] It exhibits therapeutic effects against intestinal round worms.[3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chapter 3 - Natural Products
Sharma S and Anand N
Pharmacognosy Journal , 25(7), 71-123 (1997)
Fungal hydroxylation of (-)-alpha-santonin
Bustos D A
Report Intl Narcotics Control Brd 03, 2, 1-6 (2012)
Parthenolide Covalently targets and inhibits focal adhesion kinase in breast cancer cells
Berdan C A, et al.
Cell Chemical Biology, 26(7), 1027-1035 (2019)
Takao Yamaura
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 131(3), 395-400 (2011-03-05)
Soon after its foundation in 1919, Nippon Shinyaku Co., Ltd began to develop the domestic production of Santonin, an anthelmintic agent, which, until then, had been totally imported from Russia. In 1927, Artemisia maritima ssp. monogyna was introduced from Europe
Seung Hyun Kim et al.
Archives of pharmacal research, 29(1), 40-45 (2006-02-24)
Several diacetoxy acetal analogues have been synthesized from santonin and assessed for their ability of inducing or enhancing the differentiation of human HL-60 leukemia cells. The compounds themselves had little effect on HL-60 cell differentiation. However, three analogues, 2a, 3a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service