Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1190

Sigma-Aldrich

Anti-SLIPR/MAGI-3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 140-160 kDa

species reactivity

rat

technique(s)

indirect immunofluorescence: 2.0 μg/mL using 293T cells transfected with recombinant Slipr.
microarray: suitable
western blot (chemiluminescent): 1.0 μg/mL using extracts of 293T cells transfected with recombinant Slipr

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

MAGI-3 possesses a guanylate-kinase-like region, six PDZ and two WW domains. This gene is located on human chromosome 1p13.
The MAGUK (Membrane Associated Guanylate kinase) family of proteins localize to regions of cell-cell contact, such as tight junctions in epithelial cells and synaptic junctions in neurons, and are believed to be involved in the assembly of multiprotein complexes via their protein-protein interaction domains.2

MAGI-3 (also termed SLIPR) was identified in a two hybrid screening as a protein interacting with the tumor suppressor PTEN,6 and Receptor Tyrosine Phosphatase β (RPTPβ).7

Specificity

Staining by immunoblotting and immunofluorescence is specifically inhibited by the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 835-850 of SLIPR (rat homolog of MAGI-3) conjugated to KLH via a N-terminal added lysine residue.

Biochem/physiol Actions

Membrane associated guanylate kinase, WW and PDZ domain containing 3 (MAGI3) serves as a scaffold protein for frizzled-4 and Ltap . It helps to control the c-Jun N-terminal kinase (JNK) signaling cascade.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome wide identification of new genes and pathways in patients with both autoimmune thyroiditis and type 1 diabetes
Tomer Y, et al.
Journal of Autoimmunity, 60, 32-39 (2015)
Junctional protein MAGI-3 interacts with receptor tyrosine phosphatase beta (RPTP beta) and tyrosine-phosphorylated proteins
Adamsky K, et al.
Journal of Cell Science, 116(7), 1279-1289 (2003)
Ryoji Yao et al.
Oncogene, 23(36), 6023-6030 (2004-06-15)
A seven-transmembrane protein, frizzled, has been implicated in a planar cell polarity (PCP) pathway as well as the canonical Wnt signaling pathway. Although both pathways require a cytoplasmic protein, dishevelled, the molecular mechanism by which frizzled regulates intracellular signaling remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service