Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0827

Sigma-Aldrich

Streptokinase C from Streptococcus sp.

buffered aqueous solution, ≥50,000 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous solution

specific activity

≥50,000 units/mg protein

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Streptokinase (SK) is commonly used as a thrombolytic agent in the therapy of ischemic stroke. This therapy carries the important risk of intracerebral hemorrhage. Streptokinase is also used in the treatment of complicated parapneumonic effusions and empyema where adverse reactions, allergic type, are rare. The product has been used to measure SK activity in brain heart infusion culture supernatant fluids of the various wild-type and mutant strains of Streptococcus dysgalactiae subsp. Equisimilis.[1]

Biochem/physiol Actions

Streptokinase C is an extracellular enzyme that is produced by various strains of β-hemolytic streptococci. It is a single-chain polypeptide that exerts its fibrinolytic action indirectly by activating the circulatory plasminogen.

Unit Definition

One unit will liquify a standard clot of fibrinogen, plasminogen, and thrombin at pH 7.5 at 37 °C in 10 min.

Physical form

Supplied as a frozen solution containing 60 mM phosphate buffer and 0.6 M glutamate. Albumin-free

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerstin Steiner et al.
Infection and immunity, 70(7), 3627-3636 (2002-06-18)
Synthesis of the plasminogen activator streptokinase (SK) by group A streptococci (GAS) has recently been shown to be subject to control by two two-component regulators, covRS (or csrRS) and fasBCA. In independent studies, response regulator CovR proved to act as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service