Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0774

Sigma-Aldrich

Streptomycin sulfate salt

suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H39N7O12 · 1.5 H2SO4
CAS Number:
Molecular Weight:
728.69
Beilstein:
3894995
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

form

powder

potency

≥720 μg per mg

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

technique(s)

cell culture | plant: suitable

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@@H](O[C@@H](C)[C@]2(O)C=O)O[C@@H]3[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]3NC(N)=N.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)[C@]5(O)C=O)O[C@@H]6[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]6NC(N)=N

InChI

1S/2C21H39N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,21+;;;/m00.../s1

InChI key

QTENRWWVYAAPBI-YZTFXSNBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Used together with penicillin and other agents to inhibit bacterial contamination in cell culture applications.

Biochem/physiol Actions

Mode of Action: Streptomycin acts by inhibiting prokaryote protein synthesis by binding to the S12 protein of the 30S ribosomal subunit, thus preventing the transition from imitation complex to chain-elongating ribosome. This causes miscoding or inhibits initiation.

Mode of Resistance: A mutation in rpsL, a gene for S12 ribosomal protein, prevents binding of streptomycin to the ribosome. An aminoglycoside phosphotransferase also inactivates streptomycin.

Antimicrobial spectrum: Streptomycin acts against gram-negative and gram-positive bacteria.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walter M Jaklitsch
Studies in mycology, 63, 1-91 (2009-10-15)
At present 75 species of Hypocrea have been identified in temperate Europe. Nineteen green-spored species and their Trichoderma asexual states are here described in detail. Extensive searches for Hypocrea teleomorphs in 14 European countries, with emphasis on Central Europe, yielded
Xue Tan et al.
Experimental eye research, 131, 77-83 (2015-01-13)
As a part of the visual cycle, all-trans-retinol (all-trans-ROL), the major form of vitamin A in circulating blood, is transported to the retinal pigment epithelium (RPE). All-trans-ROL is essential for normal retina function. However, recent researches have shown that excessive
Annalisa Milano et al.
Journal of the American College of Cardiology, 64(8), 745-756 (2014-08-26)
Familial forms of primary sinus bradycardia have sometimes been attributed to mutations in HCN4, SCN5A, and ANK2. In these studies, no structural cardiac alterations were reported in mutation carriers. However, a cluster of reports in the literature describe patients presenting
Ming Soon Cheng et al.
Biosensors & bioelectronics, 70, 74-80 (2015-03-22)
We describe an impedimetric cell-based biosensor constructed from poly-l-lysine (PLL)-modified screen-printed carbon electrode for real-time monitoring of dengue virus (DENV) infection of surface-immobilized baby hamster kidney (BHK-21) fibroblast cells. Cytopathic effects (CPE) induced by DENV-2 New Guinea C strain (including
Abderrazak Kitsy et al.
PloS one, 9(7), e102615-e102615 (2014-07-23)
The essential branched-chain amino acids (BCAA), leucine, valine and isoleucine, are traditionally associated with skeletal muscle growth and maintenance, energy production, and generation of neurotransmitter and gluconeogenic precursors. Recent evidence from human and animal model studies has established an additional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service