Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S014

Sigma-Aldrich

1-(3-Chlorophenyl)piperazine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClN2·2 HCl
CAS Number:
Molecular Weight:
269.60
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

SMILES string

Cl.Cl.Clc1cccc(c1)N2CCNCC2

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel F Manvich et al.
The Journal of pharmacology and experimental therapeutics, 341(2), 424-434 (2012-02-14)
Accumulating evidence indicates that the serotonin system modulates the behavioral and neurochemical effects of cocaine, but the receptor subtypes mediating these effects remain unknown. Recent studies have demonstrated that pharmacological activation of the serotonin 2C receptor (5-HT(2C)R) attenuates the behavioral
T S Gan'shina et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(8), 17-22 (2012-01-12)
Experiments on rats showed that 2-ethyl-6-methyl-3-hydroxypyridine hemisuccinate increases cerebral blood flow in the system of carotid arteries both in intact animals and under conditions of global transient ischemia. In combination with tropoxin, 2-ethyl-6-methyl-3-hydroxypyridine hemisuccinate enhances the blood flow in the
Karen A Moxon et al.
Experimental neurology, 240, 17-27 (2012-11-20)
Cortical reorganization or expansion of the intact cortical regions into the deafferented cortex after complete spinal transection in neonatally spinalized rats was shown to be essential for increases in weight-supported stepping at adulthood. The novel somatotopic organization identified in these
Carl L Faingold et al.
Brain research, 1418, 104-110 (2011-09-17)
DBA mice are models of sudden unexpected death in epilepsy (SUDEP) that exhibit audiogenic generalized convulsive seizures (GCS), ending in death due to respiratory arrest (RA). Serotonin (5-HT) normally enhances respiration in response to elevated CO(2) levels, which occur during
A A Gorbunov et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(8), 23-27 (2012-01-12)
The results of experiments on narcotized rats showed that tropoxin substantially reduces the constrictor reactions of cerebral blood vessels to meta-chlorophenylpiperazine, while not increasing the blood flow in the carotid system of either intact rats or animals with model ischemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service