Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RY865

Sigma-Aldrich

Reactive Yellow 86−Agarose

prepacked column (of R2382)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
13111000

description

Includes 5 columns, 2.5ml each

form

prepacked column (of R2382)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Kit Components Also Available Separately

Product No.
Description
SDS

  • Reactive Yellow 86−Agarose, saline suspension 5 column


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service