RT6011-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
T-60-11, non aerosol tight, without adapter, without lid, to hold, 60 x 1.5/2.0 mL tubes
Select a Size
$5,130.00
Select a Size
About This Item
$5,130.00
Recommended Products
feature
to hold 60 x 1.5/2.0 mL tubes
drum rotor (microtubes)
non aerosol tight
without adapter
without lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022638505 (NA)
Eppendorf® 5804730003 (EU)
parameter
16,400 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)