Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

RNB600

Sigma-Aldrich

GenElute Plasma/Serum RNA Purification Midi Kit

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600
NACRES:
NA.52

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

ambient

storage temp.

room temp

General description

Sigma′s GenElute Plasma/Serum RNA Purification Midi Kit provides a fast, reliable, reproducible and simple procedure for isolating circulating RNA and exosomal RNA from small plasma/serum inputs ranging from 250 μL and up to 1.5 mL, with various kit formats addressing different plasma/serum input volumes. Purification is based on spin column chromatography that uses Sigma′s proprietary resin separation matrix. The kit is designed to isolate all sizes of circulating RNA, including microRNA, as well as all sizes of exosomal RNA. Sigma′s GenElute Plasma/Serum RNA Purification Midi Kit provides a clear advantage over other available kits in that they do not require phenol/chloroform or any protease treatments. RNA can be isolated from either fresh or frozen samples using these kits. Moreover, the kits allow the user to elute into a flexible elution volume ranging from 10 μL to 100 μL. Typical yields of free-circulating and exosomal RNA vary depending on the input sample, as the amount of RNA present in plasma and serum will depend upon the health status of the individual. Normally, the RNA yield from plasma or serum RNA is highly variable (ranging from 1 to 100 ng/mL).

  • Isolate all sizes of circulating and exosomal RNA, including microRNA
  • Versatile plasma/serum input ranging from 250 μL to 1.5 mL
  • No phenol extractions
  • No carrier RNA
  • Concentrate circulating RNA and Exosomal RNA into a flexible elution volume ranging from 50 μL to 100 μL
  • Purify high-quality RNA in 35-40 minutes

Application

  • Reverse transcription qPCR
  • Reverse transcription PCR
  • Northern blotting
  • RNase protection and primer extension
  • Expression array assays
  • miRNA array
  • RNA-Seq
  • Nanostring
  • Fluidigm
  • Droplet/Digital PCR

Unit Definition

Preps

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Jing Yue et al.
Oncology letters, 16(4), 4455-4461 (2018-09-15)
Gastric cancer is an intractable disease with a poor prognosis and limited treatment options. Its treatment remains a major clinical challenge worldwide. Ephrin-B2 is upregulated and involved in tumor growth in various types of cancer. However, the association between ephrin-B2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service