Key Documents
RFA4530113-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
Select a Size
1 EA
$1,760.00
$1,760.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$1,760.00
About This Item
UNSPSC Code:
41103909
$1,760.00
Please contact Customer Service for Availability
feature
lid
to hold 30 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022654047 (NA)
Eppendorf® 5427753001 (EU)
parameter
20,817 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Features and Benefits
  • The rotor features aerosol tight quick lock rotor lid made of aluminum for use with hazardous sample
  • Fixed angle rotor holds sample at 45°
  • Compatible with 5430/5430R centrifuges
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)