RF454811-EP
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-48-11, non aerosol tight, to hold, 48 x 1.5/2.0 mL reaction vessels
Select a Size
$1,460.00
Select a Size
About This Item
$1,460.00
Recommended Products
feature
to hold 48 x 1.5/2.0 mL reaction vessels
F-45-48-11, non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822004 (North America)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)