Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RES6008H-A7

SAFC

HEPES, Hemisodium Salt

Synonym(s):

HEPES hemisodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid hemisodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) hemisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17.5N2O4S· 0.5Na
CAS Number:
Molecular Weight:
249.30
UNSPSC Code:
12352200
NACRES:
NA.21

biological source

synthetic

Assay

≥96.0% NaOH basis (Reverse titration)

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial; tested

useful pH range

6.8-8.2

pKa 

7.5

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

[Na+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

Application

HEPES Hemisodium is a biological buffer often referred to as a “Good′s” buffer. As the hemisodium salt, this buffer can be used directly to prepare a working buffer solution of HEPES. This biological buffer formulation is ideal since the pKa of the buffer is 7.31 which corresponds to the physiological pH of 7.36. HEPES Hemisodium is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

HEPES Hemisodium can be used in cell culture media, biopharmaceutical buffer formulations (both upstream and downstream) and diagnostic reagent formulations.

Packaging

Product is available in the following package sizes:
RES6008H-A701X: 100 gm container
RES6008H-A702X: 1 kg container
RES6008H-A704X: 10 kg container

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service