Skip to Content
MilliporeSigma
All Photos(1)

Documents

RAB1320

Sigma-Aldrich

Human Cathepsin Z ELISA

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

species reactivity

human

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTSZ(1522)

General description

Cathepsins are normally localized in lysosomes of almost all mammalian cells, but under certain conditions they can be secreted from the cells that take part in local proteolysis. Cathepsin Z is a cysteine protease, predominantly expressed in immune cells including monocytes, macrophages or dendritic cells.
This ELISA antibody pair detects Human Cathepsin Z (CTSZ/Cathepsin X/Cathepsin P)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Cathepsins are lysosomal proteases that play an important role in the intracellular degradation of exogenous and endogenous proteins, activation of enzyme precursors, and tumor invasion and metastasis. Cathepsin Z is known to be associated with the pathogenesis of cancer and promotes the development and proliferation of tumor cells. Cathepsin Z mediates the process of proliferation, migration, maturation, adhesion, signal transduction and phagocytosis of immune cells.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cysteine cathepsins B and X promote epithelial-mesenchymal transition of tumor cells.
Mitrovic A, et al.
European Journal of Cell Biology, 96(6), 622-631 (2017)
Localization and activity of various lysosomal proteases in Leishmania amazonensis-infected macrophages.
Prina E R I C, et al.
Infection and Immunity, 58(6), 1730-1737 (1990)
Dysregulation of apoptotic signaling pathways by interaction of RPLP0 and cathepsin X/Z in gastric cancer.
Teller A, et al.
Pathology Research and Practice, 211(1), 62-70 (2015)
Cysteine cathepsins and the cutting edge of cancer invasion.
Gocheva V and Joyce J A
Cell Cycle, 6(1), 60-64 (2007)
L Polgár et al.
The Journal of biological chemistry, 262(30), 14448-14453 (1987-10-25)
Negatively charged reactants are sensitive reactivity probes of the active site of cysteine proteases (Halász, P., and Polgár, L. (1977) Eur. J. Biochem. 79, 491-494). Thus, the thiolate-imidazolium ion pair of papain reacts at an enhanced rate with iodoacetate due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service