RAB1228
Feline CXCL1 ELISA
for serum, plasma and cell culture supernatants
Synonym(s):
C-X-C motif chemokine 1, Growth-regulated alpha protein, Platelet-derived growth factor-inducible protein KC, Secretory protein N51
About This Item
species reactivity
feline
technique(s)
ELISA: suitable
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
General description
Application
Please refer to the attached Protocolfor details.
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service