Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB1053

Sigma-Aldrich

Human GOLM1/GP-73 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$795.00

$795.00


Please contact Customer Service for Availability


Select a Size

Change View
96 WELLS
$795.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$795.00


Please contact Customer Service for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum sample(s)
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.5 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GOLM1(51280)

General description

This ELISA antibody pair detects Human GOLM1 (GP73)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed K Shaker et al.
Clinical laboratory, 66(8) (2020-08-11)
Early detection of hepatocellular carcinoma is very important in the treatment which is feasible. Alpha-fetoprotein plus ultrasound in surveillance programs is controversial. GP73 is a protein. Golgi increase significantly in the sera of patients with hepatitis B virus and HCV-related
Nikolaos K Gatselis et al.
World journal of gastroenterology, 26(34), 5130-5145 (2020-09-29)
Reliable biomarkers of cirrhosis, hepatocellular carcinoma (HCC), or progression of chronic liver diseases are missing. In this context, Golgi protein-73 (GP73) also called Golgi phosphoprotein-2, was originally defined as a resident Golgi type II transmembrane protein expressed in epithelial cells.
Marwa M Esawy et al.
Scandinavian journal of clinical and laboratory investigation, 79(7), 491-495 (2019-09-10)
GP73 is a transmembrane glycoprotein that increases in viral and non-viral liver diseases, especially in hepatocellular carcinoma. This study aims to evaluate the effect of sample type and storage conditions on GP73 concentration. Twenty subjects were enrolled in this study.
Yiming Liu et al.
Journal of cellular and molecular medicine, 23(4), 2399-2409 (2019-01-25)
Golgi phosphoprotein 73 (GP73) has been regarded as a novel serum biomarker for the diagnosis of hepatocellular carcinoma (HCC) in recent years. It has been reported that the upregulation of GP73 may promote the carcinogenesis and metastasis of HCC; however
Yun Wu et al.
Clinica chimica acta; international journal of clinical chemistry, 482, 209-214 (2018-04-10)
Development of a new method to simultaneously detect Alpha-fetoprotein (AFP) and Golgi protein 73 (GP73) from peripheral blood. Anti human AFP and GP73 monoclonal antibodies was used to develop a sandwich immunity reaction using xMAP technology for the simultaneous detection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service