Skip to Content
MilliporeSigma
All Photos(1)

Documents

RAB0455

Sigma-Aldrich

Neurokinin-A EIA Kit

for serum, plasma, culture supernatant and cell lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat, human, mouse

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TAC1(6863)
mouse ... TAC1(21333)
rat ... TAC1(24806)

General description

The Neurokinin A Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Neurokinin A peptide based on the principle of Competitive Enzyme Immunoassay.

Immunogen

Neurokinin-A, synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service