Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0452

Sigma-Aldrich

Cell-Based ELISA for detecting phospho-STAT6 (pTyr641) in cultured cell lines

96 assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

usage

96 assays

species reactivity

human, mouse

technique(s)

ELISA: suitable
capture ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT6(6778)
mouse ... STAT6(20852)

General description

The Cell-Based Stat 6 (pTyr641) ELISA kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cells. It can be used for measuring the relative amount of Stat 6 (pTyr641) phosphorylation and screening the effects of various treatments, inhibitors (such as siRNA or chemicals), or activators in cultured human and mouse cell lines.

Immunogen

Stat6 (pTyr641) synthetic phosphopeptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABBLOCK5x Blocking Solution (Item F)SDS

  • RABFIX1Fixing Solution (Item D)SDS

  • RABHRP1HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1)SDS

  • RABHRP2HRP-conjugated Anti-Mouse IgG Concentrate (Item I2)SDS

  • RABPLATE1Uncoated MicroplateSDS

  • RABQUENCHQuenching Solution for Cell-based ELISA Assay (Item E)SDS

  • RABSTOP1Stop SolutionSDS

  • RABTMB1TMB Substrate Reagent (Item J)SDS

  • RABWASH120X Wash Buffer Concentrate ASDS

  • RABWASH220X Wash Buffer Concentrate BSDS

See All (10)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service