Key Documents
RA2DWPAT-EP
Rotor for Eppendorf® 5810/5810R Centrifuges
A-2-DWP-AT, aerosol-tight
Select a Size
1 EA
$6,520.00
$6,520.00
Estimated to ship onApril 29, 2025Details
Select a Size
Change View
1 EA
$6,520.00
About This Item
UNSPSC Code:
41103909
$6,520.00
Estimated to ship onApril 29, 2025Details
feature
to hold 2 x 1 filtration plate kits
to hold 2 x 2 DWP
to hold 2 x 6 MTP
A-2-DWP-AT, aerosol-tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022639901
parameter
4,500 max. RCF (3,486 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5810/5810R
General description
Rotor for Eppendorf(R) 5418/5418R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)