Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R9404

Sigma-Aldrich

Anti-RhoA antibody produced in rabbit

~2 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-p21 Rho

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~21 kDa

species reactivity

monkey, mouse, human

concentration

~2 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK293 cells expressing human RhoA/ Hela cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOA(387)

General description

RhoA (ras homolog family member A) belongs to the Rho GTPase family of proteins. It is located on chromosome 3p21.31.

Specificity

Anti-RhoA recognizes human RhoA.

Immunogen

synthetic peptide corresponding to amino acids 177-189 located near the C-terminus of human RhoA.

Biochem/physiol Actions

In multiciliated epithelial cells, RhoA (ras homolog family member A) controls the actin network dynamics. It also modulates membrane protrusion and contractility in the cell body. It is one of the essential factor for Ras transformation. It is associated with cancer metastasis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in "frost-free" freezer, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours. For continuous use, store at 2-8°C for up to one month.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GPX1 (Glutathione Peroxidase 1)
Kulak M V and ,Weigel R J
Atlas of Genetics and Cytogenetics in Oncology and Haematology, 18(7) (2014)
Activation of Rac1, RhoA, and mitogen-activated protein kinases is required for Ras transformation
Khosravi-Far R, et al.
Molecular and Cellular Biology, 15(11), 6443-6453 (1995)
Chia-Hsin Chan et al.
Nature cell biology, 12(5), 457-467 (2010-04-13)
The RhoA GTPase is crucial in numerous biological functions and is linked to cancer metastasis. However, the understanding of the molecular mechanism responsible for RhoA transcription is still very limited. Here we show that RhoA transcription is orchestrated by the
Bruno Souza Bonifácio et al.
The Journal of eukaryotic microbiology, 69(3), e12903-e12903 (2022-03-14)
Cellular invasion by Trypanosoma cruzi metacyclic trypomastigotes (MTs) or tissue culture trypomastigotes (TCTs) is a complex process involving host-parasite cellular and molecular interactions. Particularly, the involvement of host cell actin cytoskeleton during trypomastigote invasion is poorly investigated, and still, the
RhoA regulates actin network dynamics during apical surface emergence in multiciliated epithelial cells
Sedzinski J, et al.
Journal of Cell Science, 130(2), 420-428 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service