Documents
R8133
Rulers for autoradiography
can be used with preflashed film and intensifying screens
About This Item
UNSPSC Code:
41105341
NACRES:
NB.31
manufacturer/tradename
Diversified Biotech ARS-150
General description
Non-radioactive phosphorescent emission intensity is adjusted to provide sharp markings on autoradiograms and subsequent photographs. Visible under normal lighting. Supplied with adhesive backing. Can be used with preflashed film and intensifying screens.
Packaging
Pkg. contains four 11 × 200 mm rulers.