Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R8019

Sigma-Aldrich

Ribonucleic acid, transfer, N-formylmethionine specific from Escherichia coli MRE 600

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106305

form

lyophilized powder

storage temp.

−20°C

Application

Ribonucleic acid, transfer, N-formylmethionine-specific may be used as a substrate to study the specifity and kinetics of natural, mutated and chemically engineered N-formylmethionine-specific E. coli aminoacyl-tRNA synthetase(s) and methionyl-tRNA formyltransferase(s) (MTF).

Quality

Acceptor Activity: ≥ 1000 picomoles methionine per A260 unit.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas E Jones et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(17), 6933-6938 (2011-04-13)
Aminoacyl-tRNA synthetases perform a critical step in translation by aminoacylating tRNAs with their cognate amino acids. Although high fidelity of aminoacyl-tRNA synthetases is often thought to be essential for cell biology, recent studies indicate that cells tolerate and may even
Diya M Abdeljabbar et al.
Journal of the American Chemical Society, 131(47), 17078-17079 (2009-11-10)
Engineered aminoacyl-tRNA synthetases have been used to enable the incorporation of many unnatural amino acids into recombinant proteins in vivo. In the majority of these studies, the engineered synthetase is harbored on a plasmid while the host retains a wild-type
V Ramesh et al.
Biochemistry, 37(45), 15925-15932 (1998-12-08)
Formylation of initiator methionyl-tRNA by methionyl-tRNA formyltransferase (MTF) is important for initiation of protein synthesis in eubacteria. The determinants for formylation are clustered mostly in the acceptor stem of the initiator tRNA. Previous studies suggested that a 16 amino acid
I Caglar Tanrikulu et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(36), 15285-15290 (2009-08-27)
Incorporation of noncanonical amino acids into cellular proteins often requires engineering new aminoacyl-tRNA synthetase activity into the cell. A screening strategy that relies on cell-surface display of reactive amino acid side-chains was used to identify a diverse set of methionyl-tRNA
M A Deniziak et al.
Acta biochimica Polonica, 48(2), 337-350 (2001-12-06)
Methionyl-tRNA synthetase (MetRS) belongs to the family of 20 enzymes essential for protein biosynthesis. It links covalently methionine with its cognate tRNA. Crystal structures solved for bacterial MetRSs have given a number of interesting insights into enzyme architecture and methionylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service