Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R7907

Sigma-Aldrich

RON (983-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDw136, MST1R, PTK8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

69-93 nmol/min·mg

mol wt

~71 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MST1R(4486)

Biochem/physiol Actions

RON is a macrophage stimulating 1 receptor c-met-related tyrosine kinase. RON receptor tyrosine kinase interacts with HYAL2 receptor protein, rendering it functionally inactive. HYAL2 is a candidate tumor-suppressor glycosylphosphatidylinositol-anchored cell-surface protein that serves as an entry receptor for jaagsiekte sheep retrovirus, a virus that causes contagious lung cancer in sheep that is morphologically similar to human bronchioloalveolar carcinoma. It was shown that RON liberated from the association with HYAL2 becomes functionally active and activates the Akt and mitogen-activated protein kinase pathways.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Wang et al.
Science (New York, N.Y.), 266(5182), 117-119 (1994-10-07)
Macrophage-stimulating protein (MSP) is a member of the hepatocyte growth factor-scatter factor (HGF-SF) family. Labeled MSP bound to Madin-Darby canine kidney (MDCK) cells transfected with complementary DNA encoding Ron, a cell membrane protein tyrosine kinase. Cross-linking of 125I-labeled MSP to
B Q Li et al.
Biochemical and biophysical research communications, 216(1), 110-118 (1995-11-02)
Macrophage-stimulating protein (MSP) is a chemotactic factor that activates the receptor tyrosine kinase RON. The involvement of Ras in MSP-induced signal transduction was investigated. Here we demonstrate that, in RON-transfected MDCK cells, an active GTP-bound form of Ras was rapidly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service