Skip to Content
MilliporeSigma
All Photos(1)

Documents

R7009

Sigma-Aldrich

Sty I from Escherichia coli strain carrying pST27

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.53

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-C/C(A,T)(T,A)GG-3′
Ligation and recutting results: After 2-10-fold Sty I overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >95% of fragments can be ligated, and >95% recut.
Heat inactivation: Inactivated at 65 °C for 15 minutes.

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SH (B3657).

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 0.1 mM EDTA, 50 mM KCl, 10 mM 2-mercaptoethanol, 0.1% gelatine (v/v), 0.01% polydocanol (v/v), 50% glycerol (v/v) at 4 °C

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Mise et al.
Gene, 33(3), 357-361 (1985-01-01)
A new restriction endonuclease, StyI, free of contaminating nuclease activities, has been isolated from Escherichia coli carrying the hsd+ miniplasmid of Salmonella typhi origin. In the presence of 10 mM Mg2+, it recognizes and cleaves a hexanucleotide sequence of 5'-C
C Kessler et al.
Gene, 47(1), 1-153 (1986-01-01)
The properties and sources of all known restriction endonucleases and methylases are listed. The enzymes are cross-indexed (Table I), classified according to their recognition sequence homologies (Table II), and characterized within Table II by the cleavage and methylation positions, the
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by
Saqib H Ansari et al.
Journal of pediatric hematology/oncology, 35(4), e153-e156 (2013-02-08)
β-thalassemia is characterized by impaired β-chain synthesis leading to ineffective erythropoiesis, severe anemia, and a need for blood transfusion. Presence of Xmn I polymorphism (-158 C-T nucleotide change) in γ-globin gene is associated with a higher fetal hemoglobin and a
Amal M El-Shehaby et al.
Scandinavian journal of clinical and laboratory investigation, 73(1), 75-81 (2012-12-04)
Left ventricular hypertrophy (LVH) is a common manifestation of cardiovascular disease and has an important prognostic value in patients with end-stage renal disease (ESRD). Vitamin D receptor (VDR) has been intensively investigated, and one of these (BsmI) already has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service