Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R6379

Sigma-Aldrich

Xho I from Xanthomonas holcicola

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

grade

Molecular Biology
for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-C/TCGAG-3′
Cutting results: a 2-10-fold Xho I overdigestion of 1 μg λ DNA substrate results in 100% cutting
Heat inactivation: Inactivated at 65 °C for 15 minutes.

Application

XhoI is a resteriction endonuclease used for molecular biology methods to cut DNA molecules at the recognition sequence 5′-C/TCGAG-3′ to generate fragments with 5′-cohesive ends.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SH (B3657).

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 0.1 mM EDTA, 100 mM NaCl, 10 mM 2-mercaptoethanol, 50% glycerol (v/v), 0.01% Polydocanol (v/v) at 4 °C

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new specific endonuclease present in Xanthomonas holcicola, Xanthomonas papavericola and Brevibacterium luteum.
T R Gingeras et al.
Journal of molecular biology, 118(1), 113-122 (1978-01-05)
H Sesaki et al.
The Journal of cell biology, 152(6), 1123-1134 (2001-03-21)
Membrane fusion plays an important role in controlling the shape, number, and distribution of mitochondria. In the yeast Saccharomyces cerevisiae, the outer membrane protein Fzo1p has been shown to mediate mitochondrial fusion. Using a novel genetic screen, we have isolated
B Haribabu et al.
The Journal of experimental medicine, 192(3), 433-438 (2000-08-10)
Leukotrienes are derived from arachidonic acid and serve as mediators of inflammation and immediate hypersensitivity. Leukotriene B(4) (LTB(4)) and leukotriene C(4) (LTC(4)) act through G protein-coupled receptors LTB(4) receptor (BLTR) and Cys-LTR, respectively. To investigate the physiological role of BLTR
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service