Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R5158

Sigma-Aldrich

RPMI-1640 Medium

HEPES modification with ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

RPMI culture medium, Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$73.70

$73.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$73.70

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

$73.70


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES and L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

pH

>7.2

components

phenol red: 0.0053 g/L
L-glutamine: 0.3 g/L
NaHCO3: 2 g/L
HEPES: 5.96 g/L
glucose: 2 g/L (Dextro)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.
This RPMI-1640 medium is supplemented with HEPES. HEPES is a Good′s physiological buffer that helps provide a more stable medium pH than bicarbonate buffers.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service